Learn More
n-Decyltriethoxysilane, 98%
CAS: 2943-73-9 | C16H36O3Si | 304.546 g/mol
Supplier: Thermo Scientific Chemicals L1696522
Description
n-Decyltriethoxysilane is used as coupling agent and as a chemical additive. It is also used as an intermediate of OLED, pharmaceuticals and in cosmetics. It is employed in many industries like adhesives, coatings, crude oil, glass fibers, filler treatment, polymer modification, printing, elastomers, sealants, textiles and thermoplastics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsn-Decyltriethoxysilane is used as coupling agent and as a chemical additive. It is also used as an intermediate of OLED, pharmaceuticals and in cosmetics. It is employed in many industries like adhesives, coatings, crude oil, glass fibers, filler treatment, polymer modification, printing, elastomers, sealants, textiles and thermoplastics.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. It is sensitive to moisture. Incompatible with oxidizing agents.
Specifications
| 150°C (8mmHg) | |
| 2943-73-9 | |
| C16H36O3Si | |
| MFCD00069132 | |
| >110°C (230°F) | |
| Not miscible or difficult to mix in water. | |
| CCCCCCCCCC[Si](OCC)(OCC)OCC | |
| 304.546 | |
| 304.55 | |
| 1.422 | |
| 0.879 |
| n-Decyltriethoxysilane | |
| 100 g | |
| CH3(CH2)9Si(OCH2CH3)3 | |
| 2413155 | |
| n-decyltriethoxysilane, decyl triethoxy silane, silane, decyltriethoxy | |
| BAAAEEDPKUHLID-UHFFFAOYSA-N | |
| decyl(triethoxy)silane | |
| 76261 | |
| 98% | |
| Moisture sensitive |
Chemical Identifiers
| 2943-73-9 | |
| 304.546 | |
| BAAAEEDPKUHLID-UHFFFAOYSA-N | |
| 76261 | |
| CCCCCCCCCC[Si](OCC)(OCC)OCC |
| C16H36O3Si | |
| MFCD00069132 | |
| n-decyltriethoxysilane, decyl triethoxy silane, silane, decyltriethoxy | |
| decyl(triethoxy)silane |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 220-940-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only