missing translation for 'onlineSavingsMsg'
Learn More

n-Decyltriethoxysilane, 98%

Catalog No. AAL1696514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1696514 25 g
AAL1696522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1696514 Supplier Thermo Scientific Chemicals Supplier No. L1696514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2943-73-9 | C16H36O3Si | 304.546 g/mol

n-Decyltriethoxysilane is used as coupling agent and as a chemical additive. It is also used as an intermediate of OLED, pharmaceuticals and in cosmetics. It is employed in many industries like adhesives, coatings, crude oil, glass fibers, filler treatment, polymer modification, printing, elastomers, sealants, textiles and thermoplastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Decyltriethoxysilane is used as coupling agent and as a chemical additive. It is also used as an intermediate of OLED, pharmaceuticals and in cosmetics. It is employed in many industries like adhesives, coatings, crude oil, glass fibers, filler treatment, polymer modification, printing, elastomers, sealants, textiles and thermoplastics.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. It is sensitive to moisture. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2943-73-9
Molecular Formula C16H36O3Si
Molecular Weight (g/mol) 304.546
MDL Number MFCD00069132
InChI Key BAAAEEDPKUHLID-UHFFFAOYSA-N
Synonym n-decyltriethoxysilane, decyl triethoxy silane, silane, decyltriethoxy
PubChem CID 76261
IUPAC Name decyl(triethoxy)silane
SMILES CCCCCCCCCC[Si](OCC)(OCC)OCC

Specifications

Boiling Point 150°C (8mmHg)
Quantity 25 g
Linear Formula CH3(CH2)9Si(OCH2CH3)3
Beilstein 2413155
Flash Point >110°C (230°F)
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 304.55
Percent Purity 98%
Refractive Index 1.422
Sensitivity Moisture sensitive
Density 0.879
Chemical Name or Material n-Decyltriethoxysilane
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 220-940-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only