missing translation for 'onlineSavingsMsg'
Learn More

n-Decyltrichlorosilane, 97%

Catalog No. AAL0591218
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0591218 50 g
AAL0591209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0591218 Supplier Thermo Scientific Chemicals Supplier No. L0591218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13829-21-5 | C10H21Cl3Si | 275.713 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13829-21-5
Molecular Formula C10H21Cl3Si
Molecular Weight (g/mol) 275.713
MDL Number MFCD00013600
InChI Key HLWCOIUDOLYBGD-UHFFFAOYSA-N
Synonym decyltrichlorosilane, trichloro decyl silane, n-decyltrichlorosilane, silane, trichlorodecyl, acmc-209chi, trichloro n-decyl silane, trichloro decyl silane #
PubChem CID 83764
IUPAC Name trichloro(decyl)silane
SMILES CCCCCCCCCC[Si](Cl)(Cl)Cl

Specifications

Boiling Point 133°C to 137°C (5mmHg)
Quantity 50 g
Linear Formula CH3(CH2)9SiCl3
UN Number UN2987
Beilstein 1749722
Flash Point >110°C (230°F)
Formula Weight 275.72
Percent Purity 97%
Refractive Index 1.452
Sensitivity Moisture sensitive
Density 1.049
Chemical Name or Material n-Decyltrichlorosilane
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
EINECSNumber 237-540-3
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only