missing translation for 'onlineSavingsMsg'
Learn More

n-Decylamine, 99%

Catalog No. AC111920100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111920100 10 g Glass bottle
AC111920500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC111920100 Supplier Thermo Scientific Chemicals Supplier No. 111920100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2016-57-1 | C10H23N | 157.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2016-57-1
Molecular Formula C10H23N
Molecular Weight (g/mol) 157.30
MDL Number MFCD00008149
InChI Key MHZGKXUYDGKKIU-UHFFFAOYSA-N
Synonym decylamine, 1-aminodecane, 1-decanamine, n-decylamine, 1-decylamine, decanamine, monodecylamine, kemamine p 190d, aminodecane, decyl amine
PubChem CID 8916
IUPAC Name decan-1-amine
SMILES CCCCCCCCCCN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 12°C to 15°C
Color Brown or Colorless to Yellow
Density 0.7870g/mL
Boiling Point 216°C to 218°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4350 to 1.437
Linear Formula CH3(CH2)9NH2
Quantity 10 g
Beilstein 04, 199
Specific Gravity 0.787
Formula Weight 157.3
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Decylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if inhaled.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT i
EINECSNumber 217-957-7

RUO – Research Use Only