Learn More
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, 98%
CAS: 15163-36-7 | C15H33NO3S | 307.49 g/mol
$107.65 - $107.65
Chemical Identifiers
| CAS | 15163-36-7 |
|---|---|
| Molecular Formula | C15H33NO3S |
| Molecular Weight (g/mol) | 307.49 |
| MDL Number | MFCD00036908 |
| InChI Key | WKALLSVICJPZTM-UHFFFAOYSA-N |
| Synonym | 3-decyldimethylammonio propane-1-sulfonate, n-decyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, sulfobetaine 10, caprylyl sulfobetaine, 3-decyldimethylammonio propanesulfonate, zwittergent 3-10 detergent, capryl sultaine, zwittergent 310, 3-decyl dimethyl ammonio propane-1-sulfonate, 3-decyldimethylamino propanesulfonic acid |
| PubChem CID | 161111 |
| IUPAC Name | 3-[decyl(dimethyl)azaniumyl]propane-1-sulfonate |
| SMILES | CCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC459680050
|
Thermo Scientific Chemicals
459680050 |
5 g |
Each for $107.65
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 15163-36-7 | |
| 307.49 | |
| WKALLSVICJPZTM-UHFFFAOYSA-N | |
| 161111 | |
| CCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O |
| C15H33NO3S | |
| MFCD00036908 | |
| 3-decyldimethylammonio propane-1-sulfonate, n-decyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, sulfobetaine 10, caprylyl sulfobetaine, 3-decyldimethylammonio propanesulfonate, zwittergent 3-10 detergent, capryl sultaine, zwittergent 310, 3-decyl dimethyl ammonio propane-1-sulfonate, 3-decyldimethylamino propanesulfonic acid | |
| 3-[decyl(dimethyl)azaniumyl]propane-1-sulfonate |
Specifications
| 15163-36-7 | |
| 2% max. | |
| 97.5% min. (HPLC) | |
| MFCD00036908 | |
| 3-decyldimethylammonio propane-1-sulfonate, n-decyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, sulfobetaine 10, caprylyl sulfobetaine, 3-decyldimethylammonio propanesulfonate, zwittergent 3-10 detergent, capryl sultaine, zwittergent 310, 3-decyl dimethyl ammonio propane-1-sulfonate, 3-decyldimethylamino propanesulfonic acid | |
| WKALLSVICJPZTM-UHFFFAOYSA-N | |
| 3-[decyl(dimethyl)azaniumyl]propane-1-sulfonate | |
| 161111 | |
| 98% |
| 229.0°C | |
| Authentic | |
| C15H33NO3S | |
| 5 g | |
| (10% in water) Clear colorless solution | |
| CCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O | |
| 307.49 | |
| 307.5 | |
| N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulfonate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective g
GHS Signal Word: Warning
RUO – Research Use Only