missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ n-Decanoylsucrose, ≥97%, Calbiochem™,

Catalog No. 2527211GM
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2527211GM 1 g Glass Bottle
2527215GM 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2527211GM Supplier MilliporeSigma™ Supplier No. 2527211GM

Chemical Identifiers

CAS 31835-06-0
Molecular Formula C22H40O12
Molecular Weight (g/mol) 496.55
InChI Key STVGXWVWPOLILC-UHFFFAOYSA-N
Synonym α-D-Glucopyranoside
PubChem CID 5102725
IUPAC Name [6-[3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methyl decanoate
SMILES CCCCCCCCCC(=O)OCC1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O

Specifications

Color White
Quantity 1 g
Packaging Glass Bottle
Percent Purity ≥97% (HPLC)