missing translation for 'onlineSavingsMsg'
Learn More

n-Decane (Decane; Decyl Hydride), Reagents

Catalog No. p-200020904
Change view
Click to view available options
Quantity:
100 mL
450 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337294 100 mL
01337293 450 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337294 Supplier Labchem Inc Supplier No. C2194070100C
Only null left

n-Decane (Decane; Decyl Hydride), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 124-18-5
Molecular Formula C10H22
Molecular Weight (g/mol) 142.29
InChI Key DIOQZVSQGTUSAI-UHFFFAOYSA-N
Synonym Decane, Methyl nonane, Decyl hydride
IUPAC Name decane
SMILES CCCCCCCCCC

Specifications

Boiling Range 174°C
Melting Point -29.7°C
Color Colorless
Boiling Point 174°C
Melting Range -29.7°C
Packaging Glass Bottle
Quantity 100 mL
Specific Gravity 0.73
Grade Reagent
Physical Form Liquid
Chemical Name or Material n-Decane (Decane, Decyl Hydride)
Show More Show Less