missing translation for 'onlineSavingsMsg'
Learn More

n-Decane, 99%, Spectrum™ Chemical

Catalog No. 18611917
Change view
Click to view available options
Quantity:
100 mL
500 mL
13 kg
150 kg
Packaging:
Amber Glass Bottle
Drum
Steel Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611917 100 mL Amber Glass Bottle
18611920 500 mL Amber Glass Bottle
18611918 13 kg Steel Pail
18611919 150 kg Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611917 Supplier Spectrum Chemical Mfg Cor Supplier No. D2098100ML
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 124-18-5

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 100 mL
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.