missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Cyclohexylmaleimide 98.0+%, TCI America™
$68.97 - $167.76
Chemical Identifiers
| CAS | 1631-25-0 |
|---|---|
| Molecular Formula | C10H13NO2 |
| Molecular Weight (g/mol) | 179.22 |
| MDL Number | MFCD00043904 |
| InChI Key | BQTPKSBXMONSJI-UHFFFAOYSA-N |
| Synonym | n-cyclohexylmaleimide, 1-cyclohexyl-1h-pyrrole-2,5-dione, maleimide, n-cyclohexyl, 1h-pyrrole-2,5-dione, 1-cyclohexyl, n-cyclohexyl maleimide, 1-cyclohexylazoline-2,5-dione, acmc-1btr9, 1h-pyrrole-2, 1-cyclohexyl, 1-cyclohexyl-pyrrole-2,5-dione |
| PubChem CID | 74203 |
| IUPAC Name | 1-cyclohexyl-2,5-dihydro-1H-pyrrole-2,5-dione |
| SMILES | O=C1C=CC(=O)N1C1CCCCC1 |
Chemical Identifiers
| 1631-25-0 | |
| 179.22 | |
| BQTPKSBXMONSJI-UHFFFAOYSA-N | |
| 74203 | |
| O=C1C=CC(=O)N1C1CCCCC1 |
| C10H13NO2 | |
| MFCD00043904 | |
| n-cyclohexylmaleimide, 1-cyclohexyl-1h-pyrrole-2,5-dione, maleimide, n-cyclohexyl, 1h-pyrrole-2,5-dione, 1-cyclohexyl, n-cyclohexyl maleimide, 1-cyclohexylazoline-2,5-dione, acmc-1btr9, 1h-pyrrole-2, 1-cyclohexyl, 1-cyclohexyl-pyrrole-2,5-dione | |
| 1-cyclohexyl-2,5-dihydro-1H-pyrrole-2,5-dione |
Specifications
| 1631-25-0 | |
| White-Yellow | |
| C10H13NO2 | |
| 25 g | |
| n-cyclohexylmaleimide, 1-cyclohexyl-1h-pyrrole-2,5-dione, maleimide, n-cyclohexyl, 1h-pyrrole-2,5-dione, 1-cyclohexyl, n-cyclohexyl maleimide, 1-cyclohexylazoline-2,5-dione, acmc-1btr9, 1h-pyrrole-2, 1-cyclohexyl, 1-cyclohexyl-pyrrole-2,5-dione | |
| O=C1C=CC(=O)N1C1CCCCC1 | |
| 179.22 | |
| 179.22 | |
| Crystalline Powder |
| 88°C | |
| 132°C | |
| MFCD00043904 | |
| 2811 | |
| BQTPKSBXMONSJI-UHFFFAOYSA-N | |
| 1-cyclohexyl-2,5-dihydro-1H-pyrrole-2,5-dione | |
| 74203 | |
| ≥98.0% (GC) | |
| N-Cyclohexylmaleimide |
Safety and Handling
EINECSNumber : (5)-5862
TSCA : Yes