missing translation for 'onlineSavingsMsg'
Learn More

N-Cyclohexylhydroxylamine hydrochloride, 98%

Catalog No. AAL0356903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0356903 1 g
AAL0356906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0356903 Supplier Thermo Scientific Chemicals Supplier No. L0356903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25100-12-3 | C6H14ClNO | 151.634 g/mol

N-Cyclohexylhydroxylamine hydrochloride is used in the preparation of 4-carbethoxy-2-cyclohexyl-5-(2H)-isoxazolone and hexahydroisoxazolequinoline analogs. It is also used in the preparation of N-cyclohexyl nitrone by reaction with aldehyde. Further, it acts as a hydroxylamination reagent for allyl esters. In addition to this, it serves as a reagent in the conversion of 1,2-dicarboxylic acids to alfa-beta-unsaturated carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Cyclohexylhydroxylamine hydrochloride is used in the preparation of 4-carbethoxy-2-cyclohexyl-5-(2H)-isoxazolone and hexahydroisoxazolequinoline analogs. It is also used in the preparation of N-cyclohexyl nitrone by reaction with aldehyde. Further, it acts as a hydroxylamination reagent for allyl esters. In addition to this, it serves as a reagent in the conversion of 1,2-dicarboxylic acids to alfa-beta-unsaturated carboxylic acids.

Solubility
Soluble in water.

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 25100-12-3
Molecular Formula C6H14ClNO
Molecular Weight (g/mol) 151.634
MDL Number MFCD00012565
InChI Key SSVAHXZUFFSFER-UHFFFAOYSA-N
Synonym n-cyclohexylhydroxylamine hydrochloride, cyclohexylhydroxylamine hydrochloride, unii-r98vsp3t5l, r98vsp3t5l, cyclohexylhydroxylamine hcl, acmc-1ch2g, n-cyclohexylhydroxylarnine hydrochloride, n-cyclohexyl hydroxylamine hydrochloride, cyclohexanamine,n-hydroxy-,hydrochloride 1:1
PubChem CID 3084432
IUPAC Name N-cyclohexylhydroxylamine;hydrochloride
SMILES C1CCC(CC1)NO.Cl

Specifications

Melting Point ∽166°C (decomposition)
Quantity 1 g
Beilstein 3672798
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 151.64
Percent Purity 98%
Chemical Name or Material N-Cyclohexylhydroxylamine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 246-612-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only