missing translation for 'onlineSavingsMsg'
Learn More

N-Cyclohexylethanolamine 98.0+%, TCI America™

Catalog No. C1082500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1082500G 500 g
C108225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1082500G Supplier TCI America Supplier No. C1082500G
Only null left

Chemical Identifiers

CAS 2842-38-8
Molecular Formula C8H17NO
Molecular Weight (g/mol) 143.23
MDL Number MFCD00021326
InChI Key MGUMZJAQENFQKN-UHFFFAOYSA-N
Synonym n-cyclohexylethanolamine, 2-cyclohexylamino ethanol, ethanol, 2-cyclohexylamino, abbomeen e-25 aerosol, cyclohexlaminoethanol, abromeen e-25, 2-cyclohexylamino ethan-1-ol, n-2-hydroxyethyl cyclohexylamine, cyclohexylamine, hydroxyethyl
PubChem CID 17827
IUPAC Name 2-(cyclohexylamino)ethanol
SMILES C1CCC(CC1)NCCO

Specifications

Color White
Quantity 500 g
Formula Weight 143.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material N-Cyclohexylethanolamine
RTECSNumber KK3530000
TSCA No