missing translation for 'onlineSavingsMsg'
Learn More

N-(Cyanomethyl)pyridinium chloride, 97%

Catalog No. p-7021830
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1432718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1432718 Supplier Thermo Scientific Chemicals Supplier No. A1432718

CAS: 17281-59-3 | C7H7ClN2 | 154.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17281-59-3
Molecular Formula C7H7ClN2
Molecular Weight (g/mol) 154.60
MDL Number MFCD00031933
InChI Key HEJOROWXIWLCMS-UHFFFAOYSA-M
Synonym 1-cyanomethyl pyridinium chloride, n-cyanomethyl pyridinium chloride, 1-cyanomethyl pyridin-1-ium chloride, acmc-1chuo, cyanomethylpyridinium chloride, n-cyanomethyl pyridiniumchloride, 2-pyridin-1-ium-1-ylacetonitrile chloride, 2-1-pyridin-1-iumyl acetonitrile chloride, 2-pyridin-1-ium-1-ylethanenitrile chloride
PubChem CID 2734116
IUPAC Name 2-pyridin-1-ium-1-ylacetonitrile;chloride
SMILES [Cl-].N#CC[N+]1=CC=CC=C1

Specifications

Melting Point 173°C to 175°C
Quantity 50 g
UN Number UN3439
Beilstein 3728392
Formula Weight 154.6
Percent Purity 97%
Chemical Name or Material N-(Cyanomethyl)pyridinium chloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 241-312-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only