missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Chlorosuccinimide 98.0+%, TCI America™
$32.72 - $130.92
Chemical Identifiers
| CAS | 128-09-6 |
|---|---|
| Molecular Formula | C4H4ClNO2 |
| Molecular Weight (g/mol) | 133.531 |
| MDL Number | MFCD00005511 |
| InChI Key | JRNVZBWKYDBUCA-UHFFFAOYSA-N |
| Synonym | n-chlorosuccinimide, succinchlorimide, chlorosuccinimide, succinochlorimide, succinic n-chloroimide, 2,5-pyrrolidinedione, 1-chloro, 1-chloro-2,5-pyrrolidinedione, succinimide, n-chloro, ncs, caswell no. 807 |
| PubChem CID | 31398 |
| ChEBI | CHEBI:53203 |
| IUPAC Name | 1-chloropyrrolidine-2,5-dione |
| SMILES | C1CC(=O)N(C1=O)Cl |
Chemical Identifiers
| 128-09-6 | |
| 133.531 | |
| JRNVZBWKYDBUCA-UHFFFAOYSA-N | |
| 31398 | |
| 1-chloropyrrolidine-2,5-dione |
| C4H4ClNO2 | |
| MFCD00005511 | |
| n-chlorosuccinimide, succinchlorimide, chlorosuccinimide, succinochlorimide, succinic n-chloroimide, 2,5-pyrrolidinedione, 1-chloro, 1-chloro-2,5-pyrrolidinedione, succinimide, n-chloro, ncs, caswell no. 807 | |
| CHEBI:53203 | |
| C1CC(=O)N(C1=O)Cl |
Specifications
| 128-09-6 | |
| White | |
| MFCD00005511 | |
| 3261 | |
| JRNVZBWKYDBUCA-UHFFFAOYSA-N | |
| 1-chloropyrrolidine-2,5-dione | |
| 31398 | |
| 133.53 | |
| Crystalline Powder |
| 148°C | |
| C4H4ClNO2 | |
| 25 g | |
| n-chlorosuccinimide, succinchlorimide, chlorosuccinimide, succinochlorimide, succinic n-chloroimide, 2,5-pyrrolidinedione, 1-chloro, 1-chloro-2,5-pyrrolidinedione, succinimide, n-chloro, ncs, caswell no. 807 | |
| C1CC(=O)N(C1=O)Cl | |
| 133.531 | |
| CHEBI:53203 | |
| ≥98.0% (T) | |
| N-Chlorosuccinimide |
Safety and Handling
EINECSNumber : (5)-0121
RTECSNumber : UY1013500
TSCA : Yes