missing translation for 'onlineSavingsMsg'
Learn More

N-Chlorosuccinimide 98.0+%, TCI America™

Catalog No. C0291500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0291500G 500 g
C029125G 25 g
C0291100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0291500G Supplier TCI America Supplier No. C0291500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 128-09-6
Molecular Formula C4H4ClNO2
Molecular Weight (g/mol) 133.531
MDL Number MFCD00005511
InChI Key JRNVZBWKYDBUCA-UHFFFAOYSA-N
Synonym n-chlorosuccinimide, succinchlorimide, chlorosuccinimide, succinochlorimide, succinic n-chloroimide, 2,5-pyrrolidinedione, 1-chloro, 1-chloro-2,5-pyrrolidinedione, succinimide, n-chloro, ncs, caswell no. 807
PubChem CID 31398
ChEBI CHEBI:53203
IUPAC Name 1-chloropyrrolidine-2,5-dione
SMILES C1CC(=O)N(C1=O)Cl

Specifications

Melting Point 148°C
Color White
Quantity 500 g
UN Number 3261
Formula Weight 133.53
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Chlorosuccinimide
EINECSNumber (5)-0121
RTECSNumber UY1013500
TSCA Yes