missing translation for 'onlineSavingsMsg'
Learn More

N-Chlorosuccinimide, 98%

CAS: 128-09-6 | C4H4ClNO2 | 133.531 g/mol

Supplier:  Thermo Scientific Chemicals A103100B

 View more versions of this product

Catalog No. AAA103100B


Only null left
Explore available promotions

Description

Description

Regioselective chlorination and oxidizing reagentN-Chlorosuccinimide is used as a chlorinating agent, mild oxidant and swimming pool disinfectant. It is used in the microdetermination of hydrazine salts by titrimetry as well as in the detection of arsenic in the +3 oxidative state. It is also employed in the production of rubber additives. Further, it is used as pharmaceutical intermediate and antibiotic drugs intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Regioselective chlorination and oxidizing reagentN-Chlorosuccinimide is used as a chlorinating agent, mild oxidant and swimming pool disinfectant. It is used in the microdetermination of hydrazine salts by titrimetry as well as in the detection of arsenic in the +3 oxidative state. It is also employed in the production of rubber additives. Further, it is used as pharmaceutical intermediate and antibiotic drugs intermediate.

Solubility
Soluble in water, alcohol, benzene, acetone and acetic acid. Slightly soluble in ether, chloroform, carbon tetrachloride and petroleum ether.

Notes
Light sensitive and moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases, amines, ammonia, iron and iron salts.
Specifications

Specifications

N-Chlorosuccinimide
146°C to 150°C
Chlorine-like
MFCD00005511
UN3261
Moisture sensitive
n-chlorosuccinimide, succinchlorimide, chlorosuccinimide, succinochlorimide, succinic n-chloroimide, 2,5-pyrrolidinedione, 1-chloro, 1-chloro-2,5-pyrrolidinedione, succinimide, n-chloro, ncs, caswell no. 807
JRNVZBWKYDBUCA-UHFFFAOYSA-N
1-chloropyrrolidine-2,5-dione
31398
133.53
128-09-6
1.65
C4H4ClNO2
1000 g
113915
14,2164
Soluble in water,alcohol,benzene,acetone and acetic acid. Slightly soluble in ether,chloroform,carbon tetrachloride and petroleum ether.
C1CC(=O)N(C1=O)Cl
133.531
CHEBI:53203
98%

Chemical Identifiers

128-09-6
133.531
JRNVZBWKYDBUCA-UHFFFAOYSA-N
31398
1-chloropyrrolidine-2,5-dione
C4H4ClNO2
MFCD00005511
n-chlorosuccinimide, succinchlorimide, chlorosuccinimide, succinochlorimide, succinic n-chloroimide, 2,5-pyrrolidinedione, 1-chloro, 1-chloro-2,5-pyrrolidinedione, succinimide, n-chloro, ncs, caswell no. 807
CHEBI:53203
C1CC(=O)N(C1=O)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302;
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P363-P501c

H302-H314-H335-H500

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 204-878-8

RTECSNumber : UY1013500

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only