missing translation for 'onlineSavingsMsg'
Learn More

N-Chlorosuccinimide, 98%

Catalog No. AAA1031018
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1031018 50 g
AAA1031030 250 g
AAA103100B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1031018 Supplier Thermo Scientific Chemicals Supplier No. A1031018
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 128-09-6 | C4H4ClNO2 | 133.531 g/mol

Regioselective chlorination and oxidizing reagentN-Chlorosuccinimide is used as a chlorinating agent, mild oxidant and swimming pool disinfectant. It is used in the microdetermination of hydrazine salts by titrimetry as well as in the detection of arsenic in the +3 oxidative state. It is also employed in the production of rubber additives. Further, it is used as pharmaceutical intermediate and antibiotic drugs intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Regioselective chlorination and oxidizing reagentN-Chlorosuccinimide is used as a chlorinating agent, mild oxidant and swimming pool disinfectant. It is used in the microdetermination of hydrazine salts by titrimetry as well as in the detection of arsenic in the +3 oxidative state. It is also employed in the production of rubber additives. Further, it is used as pharmaceutical intermediate and antibiotic drugs intermediate.

Solubility
Soluble in water, alcohol, benzene, acetone and acetic acid. Slightly soluble in ether, chloroform, carbon tetrachloride and petroleum ether.

Notes
Light sensitive and moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases, amines, ammonia, iron and iron salts.

Chemical Identifiers

CAS 128-09-6
Molecular Formula C4H4ClNO2
Molecular Weight (g/mol) 133.531
MDL Number MFCD00005511
InChI Key JRNVZBWKYDBUCA-UHFFFAOYSA-N
Synonym n-chlorosuccinimide, succinchlorimide, chlorosuccinimide, succinochlorimide, succinic n-chloroimide, 2,5-pyrrolidinedione, 1-chloro, 1-chloro-2,5-pyrrolidinedione, succinimide, n-chloro, ncs, caswell no. 807
PubChem CID 31398
ChEBI CHEBI:53203
IUPAC Name 1-chloropyrrolidine-2,5-dione
SMILES C1CC(=O)N(C1=O)Cl

Specifications

Melting Point 146°C to 150°C
Density 1.65 g/mL
Assay Percent Range 98%
Quantity 50 g
Beilstein 113915
Sensitivity Moisture sensitive
Solubility Information Soluble in water,alcohol,benzene,acetone and acetic acid. Slightly soluble in ether,chloroform,carbon tetrachloride and petroleum ether.
Formula Weight 133.53
Chemical Name or Material N-Chlorosuccinimide, 98%
Hazard Category H302-H314-H335-H500
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 204-878-8
RTECSNumber UY1013500
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only