missing translation for 'onlineSavingsMsg'
Learn More

N-Chlorophthalimide 95.0+%, TCI America™

Catalog No. p-7124617
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C080225G 25 g
C0802500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C080225G Supplier TCI America Supplier No. C080225G

Chemical Identifiers

CAS 3481-09-2
Molecular Formula C8H4ClNO2
Molecular Weight (g/mol) 181.58
MDL Number MFCD00023027
InChI Key WDRFYIPWHMGQPN-UHFFFAOYSA-N
Synonym n-chlorophthalimide, 2-chloroisoindoline-1,3-dione, phthalimide, n-chloro, 1h-isoindole-1,3 2h-dione, 2-chloro, n-chlorphthalimid, n-chlorophtalimide, phthalimide chloride, acmc-1aglo, n-chloro phthalamide
PubChem CID 18997
IUPAC Name 2-chloroisoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)Cl

Specifications

Melting Point 186°C
Color White
Quantity 25 g
UN Number 1759
Formula Weight 181.58
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Chlorophthalimide