missing translation for 'onlineSavingsMsg'
Learn More

N-Chloroacetyl-L-leucine 99.0+%, TCI America™

Catalog No. p-7124216
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C01011G 1 g
C010125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C01011G Supplier TCI America Supplier No. C01011G

Chemical Identifiers

CAS 688-12-0
Molecular Formula C8H14ClNO3
Molecular Weight (g/mol) 207.65
MDL Number MFCD00018912
InChI Key VDUNMYRYEYROFL-UHFFFAOYNA-N
PubChem CID 2795110
IUPAC Name 2-(2-chloroacetamido)-4-methylpentanoic acid
SMILES CC(C)CC(NC(=O)CCl)C(O)=O

Specifications

Melting Point 134°C
Color White
Quantity 1 g
Formula Weight 207.65
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Chloroacetyl-L-leucine