missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzyloxyglycine, 98.5%

Catalog No. p-83011
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108350250 25 g Plastic Bottle
AC108351000 100 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC108350250 Supplier Thermo Scientific Chemicals Supplier No. 108350250
Only null left

CAS: 1138-80-3 | C10H11NO4 | 209.201 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1138-80-3
MDL Number MFCD00002691
InChI Key CJUMAFVKTCBCJK-UHFFFAOYSA-N
Synonym n-carbobenzyloxyglycine, n-cbz-glycine, z-gly-oh, carbobenzyloxyglycine, carbobenzoxyglycine, n-benzyloxycarbonylglycine, n-carbobenzoxyglycine, cbz-gly-oh, cbz gly, benzyloxycarbonylglycine
PubChem CID 14349
ChEBI CHEBI:16532
IUPAC Name 2-(phenylmethoxycarbonylamino)acetic acid
SMILES C1=CC=C(C=C1)COC(=O)NCC(=O)O

Specifications

Melting Point 117.0°C to 121.0°C
CAS Min % 98.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5%
Packaging Plastic Bottle
Molecular Formula C10H11NO4
Linear Formula C6H5CH2O2CNHCH2CO2H
Quantity 25 g
Molecular Weight (g/mol) 209.201
Formula Weight 209.2
Percent Purity 98.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material N-Carbobenzyloxyglycine, 98.5%
Show More Show Less
EINECSNumber 214-516-0
RTECSNumber MB9129000
TSCA TSCA

RUO – Research Use Only