missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Carbobenzyloxy-L-phenylalanine, 99+%

Catalog No. AC275960050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC275960050 5 g Glass bottle
AC275960250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC275960050 Supplier Thermo Scientific Chemicals Supplier No. 275960050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1161-13-3 | C17H17NO4 | 299.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1161-13-3
Molecular Formula C17H17NO4
Molecular Weight (g/mol) 299.33
MDL Number MFCD00020418
InChI Key RRONHWAVOYADJL-HNNXBMFYSA-N
Synonym z-phe-oh, n-cbz-l-phenylalanine, carbobenzoxyphenylalanine, cbz-phe-oh, n-carbobenzoxy-l-phenylalanine, carbobenzoxy-l-phenylalanine, z-l-phenylalanine, benzyloxycarbonyl phenylalanine, n-carbobenzyloxy-l-phenylalanine
PubChem CID 70878
IUPAC Name (2S)-3-phenyl-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES OC(=O)[C@H](CC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 85°C to 88°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH2CH(NHCO2CH2C6H5)CO2H
Quantity 5 g
Specific Rotation + 5
Formula Weight 299.32
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material N-Carbobenzyloxy-L-phenylalanine, 99+%
Show More Show Less
EINECSNumber 214-599-3
RTECSNumber AY4343000

RUO – Research Use Only