missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Carbobenzyloxy-L-aspartic acid, 98%

Catalog No. AC172051000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC172051000 100 g
AC172050250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC172051000 Supplier Thermo Scientific Chemicals Supplier No. 172051000

CAS: 1152-61-0 | C12H13NO6 | 267.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1152-61-0
Molecular Formula C12H13NO6
Molecular Weight (g/mol) 267.23
MDL Number MFCD00002719
InChI Key XYXYXSKSTZAEJW-VIFPVBQESA-N
Synonym z-asp-oh, n-carbobenzyloxy-l-aspartic acid, cbz-asp-oh, n-carbobenzoxy-l-aspartic acid, n-cbz-l-aspartic acid, z-l-aspartic acid, carbobenzoxy-l-aspartic acid, n-z-l-aspartic acid, n-benzyloxycarbonylaspartic acid, z-l-asp-oh
PubChem CID 2723942
IUPAC Name (2S)-2-(phenylmethoxycarbonylamino)butanedioic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CC(=O)O)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 117°C to 119°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HO2CCH2CH(NHCO2CH2C6H5)CO2H
Quantity 100 g
Specific Rotation Condition +10.50° (20°C c=1,AcOH)
Specific Rotation + 10.5
Solubility Information Solubility in water: .. Other solubilities: soluble in most common organic solvents
Formula Weight 267.23
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material N-Carbobenzyloxy-L-aspartic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-568-4

RUO – Research Use Only