missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Carbobenzyloxy-L-alanine, 98%

Catalog No. p-3757185
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169000050 5 g Glass bottle
AC169000250 25 g Glass bottle
AC169001000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC169000050 Supplier Thermo Scientific Chemicals Supplier No. 169000050

CAS: 1142-20-7 | C11H13NO4 | 223.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1142-20-7
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
MDL Number MFCD00002640
InChI Key TYRGLVWXHJRKMT-QMMMGPOBSA-N
Synonym n-carbobenzyloxy-l-alanine, z-ala-oh, n-cbz-l-alanine, z-l-alanine, n-benzyloxycarbonyl-l-alanine, cbz-l-alanine, cbz-ala-oh, carbobenzyloxy-l-alanine, benzyloxycarbonyl-l-alanine, carbobenzoxy-l-alanine
PubChem CID 736104
IUPAC Name (2S)-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 82°C to 84°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3CH(NHCO2CH2C6H5)CO2H
Quantity 5 g
Specific Rotation -15
Solubility Information Solubility in water: .. Other solubilities: soluble in most common organic solvents
Formula Weight 223.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Carbobenzyloxy-L-alanine
Show More Show Less
EINECSNumber 214-532-8

RUO – Research Use Only