missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxyglycine 98.0+%, TCI America™

Catalog No. C057525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C057525G 25 g
C0575500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C057525G Supplier TCI America Supplier No. C057525G
Only null left

Chemical Identifiers

CAS 1138-80-3
Molecular Formula C10H11NO4
Molecular Weight (g/mol) 209.201
MDL Number MFCD00002691
InChI Key CJUMAFVKTCBCJK-UHFFFAOYSA-N
Synonym n-carbobenzyloxyglycine, n-cbz-glycine, z-gly-oh, carbobenzyloxyglycine, carbobenzoxyglycine, n-benzyloxycarbonylglycine, n-carbobenzoxyglycine, cbz-gly-oh, cbz gly, benzyloxycarbonylglycine
PubChem CID 14349
ChEBI CHEBI:16532
IUPAC Name 2-(phenylmethoxycarbonylamino)acetic acid
SMILES C1=CC=C(C=C1)COC(=O)NCC(=O)O

Specifications

Melting Point 120°C
Color White
Quantity 25 g
Formula Weight 209.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxyglycine
EINECSNumber (3)-4607
RTECSNumber MB9129000
TSCA Yes