missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-N-methylglycine 97.0+%, TCI America™
$65.24 - $216.24
Chemical Identifiers
| CAS | 39608-31-6 |
|---|---|
| Molecular Formula | C11H13NO4 |
| Molecular Weight (g/mol) | 223.228 |
| MDL Number | MFCD00021747 |
| InChI Key | CBWFTZNMONHKNZ-UHFFFAOYSA-N |
| Synonym | z-sar-oh, cbz-sar-oh, n-benzyloxycarbonyl sarcosine, carbobenzoxy sarcosine, z-sarcosine, glycine, n-methyl-n-phenylmethoxy carbonyl, n-cbz-sarcosine, 2-benzyloxy carbonyl methyl amino acetic acid, n-carbobenzoxy sarcosine, benzyloxycarbonyl sarcosine |
| PubChem CID | 97053 |
| IUPAC Name | 2-[methyl(phenylmethoxycarbonyl)amino]acetic acid |
| SMILES | CN(CC(=O)O)C(=O)OCC1=CC=CC=C1 |
Chemical Identifiers
| 39608-31-6 | |
| 223.228 | |
| CBWFTZNMONHKNZ-UHFFFAOYSA-N | |
| 97053 | |
| CN(CC(=O)O)C(=O)OCC1=CC=CC=C1 |
| C11H13NO4 | |
| MFCD00021747 | |
| z-sar-oh, cbz-sar-oh, n-benzyloxycarbonyl sarcosine, carbobenzoxy sarcosine, z-sarcosine, glycine, n-methyl-n-phenylmethoxy carbonyl, n-cbz-sarcosine, 2-benzyloxy carbonyl methyl amino acetic acid, n-carbobenzoxy sarcosine, benzyloxycarbonyl sarcosine | |
| 2-[methyl(phenylmethoxycarbonyl)amino]acetic acid |
Specifications
| 39608-31-6 | |
| C11H13NO4 | |
| z-sar-oh, cbz-sar-oh, n-benzyloxycarbonyl sarcosine, carbobenzoxy sarcosine, z-sarcosine, glycine, n-methyl-n-phenylmethoxy carbonyl, n-cbz-sarcosine, 2-benzyloxy carbonyl methyl amino acetic acid, n-carbobenzoxy sarcosine, benzyloxycarbonyl sarcosine | |
| CN(CC(=O)O)C(=O)OCC1=CC=CC=C1 | |
| 5 g | |
| 97053 | |
| ≥97.0% (HPLC) | |
| N-Carbobenzoxy-N-methylglycine |
| Yellow | |
| MFCD00021747 | |
| CBWFTZNMONHKNZ-UHFFFAOYSA-N | |
| 2-[methyl(phenylmethoxycarbonyl)amino]acetic acid | |
| 223.228 | |
| 223.23 | |
| Liquid |
Safety and Handling
TSCA : Yes