missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-N-methylglycine 97.0+%, TCI America™

Catalog No. C28485G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28485G 5 g
C284825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28485G Supplier TCI America Supplier No. C28485G

Chemical Identifiers

CAS 39608-31-6
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.228
MDL Number MFCD00021747
InChI Key CBWFTZNMONHKNZ-UHFFFAOYSA-N
Synonym z-sar-oh, cbz-sar-oh, n-benzyloxycarbonyl sarcosine, carbobenzoxy sarcosine, z-sarcosine, glycine, n-methyl-n-phenylmethoxy carbonyl, n-cbz-sarcosine, 2-benzyloxy carbonyl methyl amino acetic acid, n-carbobenzoxy sarcosine, benzyloxycarbonyl sarcosine
PubChem CID 97053
IUPAC Name 2-[methyl(phenylmethoxycarbonyl)amino]acetic acid
SMILES CN(CC(=O)O)C(=O)OCC1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 223.23
Percent Purity ≥97.0% (HPLC)
Physical Form Liquid
Chemical Name or Material N-Carbobenzoxy-N-methylglycine