missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-L-tryptophan 98.0+%, TCI America™
Supplier: TCI America C063825G
Specifications
| N-Carbobenzoxy-L-tryptophan | |
| 125°C | |
| C19H18N2O4 | |
| 25 g | |
| AHYFYYVVAXRMKB-KRWDZBQOSA-N | |
| (2S)-2-{[(benzyloxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoic acid | |
| 101176 | |
| ≥98.0% (HPLC,T) |
| 7432-21-5 | |
| White-Yellow | |
| MFCD00065700 | |
| z-trp-oh, n-cbz-l-tryptophan, z-l-trp-oh, benzyloxycarbonyltryptophan, l-tryptophan, n-phenylmethoxy carbonyl, n-carbobenzoxyltryptophan, n-carbobenzoxy-l-tryptophan, cbz-l-trp-oh, n-benzyloxycarbonyl-l-tryptophan, nalpha-carbobenzyloxy-l-tryptophan | |
| OC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)OCC1=CC=CC=C1 | |
| 338.36 | |
| 338.36 | |
| Crystalline Powder |
Chemical Identifiers
| 7432-21-5 | |
| 338.36 | |
| AHYFYYVVAXRMKB-KRWDZBQOSA-N | |
| 101176 | |
| OC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)OCC1=CC=CC=C1 |
| C19H18N2O4 | |
| MFCD00065700 | |
| z-trp-oh, n-cbz-l-tryptophan, z-l-trp-oh, benzyloxycarbonyltryptophan, l-tryptophan, n-phenylmethoxy carbonyl, n-carbobenzoxyltryptophan, n-carbobenzoxy-l-tryptophan, cbz-l-trp-oh, n-benzyloxycarbonyl-l-tryptophan, nalpha-carbobenzyloxy-l-tryptophan | |
| (2S)-2-{[(benzyloxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoic acid |
Safety and Handling
TSCA : No