missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-L-pyroglutamic Acid 98.0+%, TCI America™

Catalog No. p-7125853
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31435G 5 g
C314325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31435G Supplier TCI America Supplier No. C31435G

Chemical Identifiers

CAS 32159-21-0
Molecular Formula C13H13NO5
Molecular Weight (g/mol) 263.25
MDL Number MFCD00037352
InChI Key VHSFUGXCSGOKJX-UHFFFAOYNA-N
Synonym z-pyr-oh, cbz-l-pyroglutamic acid, z-l-pyroglutamic acid, benzyloxycarbonyl-l-pyroglutamic acid, s-5-oxo-pyrrolidine-1,2-dicarboxylic acid 1-benzyl ester, s-1-benzyloxy carbonyl-5-oxopyrrolidine-2-carboxylic acid, s-1-benzyloxycarbonyl-5-oxopyrrolidine-2-carboxylic acid, 2s-5-oxo-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid, z-pglu-oh
PubChem CID 637648
IUPAC Name 1-[(benzyloxy)carbonyl]-5-oxopyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCC(=O)N1C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 141°C
Color White
Quantity 5 g
Formula Weight 263.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-pyroglutamic Acid
TSCA No
Recommended Storage Freezer (-20 °C)