missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-L-isoleucine Dicyclohexylammonium Salt 98.0+%, TCI America™

Catalog No. C07591G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C07591G Supplier TCI America Supplier No. C07591G
Only null left

Chemical Identifiers

CAS 26699-00-3
Molecular Formula C26H42N2O4
Molecular Weight (g/mol) 446.63
MDL Number MFCD00043243
InChI Key QWMHUFMEYKIYPC-QUTFAALHNA-N
Synonym cbz-l-isoleucine dicyclohexylamine salt, z-ile-oh dicyclohexylammonium salt, cbz-ile-oh.dcha, n-benzyloxy carbonyl-l-isoleucine, compound with dicyclohexylamine 1:1, cbz-isoleucine.dcha, cbz-ile-oh??dcha, pubchem18961, z-l-isoleucine dicyclohexylammonium salt, cbz-l-isoleucine dicyclohexylammonium salt, n-alpha-benzyloxycarbonyl-l-isoleucine dicyclohexylamine
PubChem CID 43835055
IUPAC Name (2S)-2-{[(benzyloxy)carbonyl]amino}-3-methylpentanoic acid; N-cyclohexylcyclohexanamine
SMILES C1CCC(CC1)NC1CCCCC1.CCC(C)[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 161°C
Color White
Quantity 1 g
Formula Weight 446.63
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-isoleucine Dicyclohexylammonium Salt