missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-L-homoserine Lactone 98.0+%, TCI America™
$52.19 - $206.90
Chemical Identifiers
| CAS | 35677-89-5 |
|---|---|
| Molecular Formula | C12H13NO4 |
| Molecular Weight (g/mol) | 235.239 |
| MDL Number | MFCD00674555 |
| InChI Key | FKWDZIFOVOUDAG-JTQLQIEISA-N |
| Synonym | N-Cbz-L-homoserine Lactone, (S)-alpha-(Carbobenzoxyamino)-gamma-butyrolactone, (S)-alpha-(Cbz-amino)-gamma-butyrolactone |
| PubChem CID | 489185 |
| IUPAC Name | benzyl N-[(3S)-2-oxooxolan-3-yl]carbamate |
| SMILES | C1COC(=O)C1NC(=O)OCC2=CC=CC=C2 |
Chemical Identifiers
| 35677-89-5 | |
| 235.239 | |
| FKWDZIFOVOUDAG-JTQLQIEISA-N | |
| 489185 | |
| C1COC(=O)C1NC(=O)OCC2=CC=CC=C2 |
| C12H13NO4 | |
| MFCD00674555 | |
| N-Cbz-L-homoserine Lactone, (S)-alpha-(Carbobenzoxyamino)-gamma-butyrolactone, (S)-alpha-(Cbz-amino)-gamma-butyrolactone | |
| benzyl N-[(3S)-2-oxooxolan-3-yl]carbamate |
Specifications
| 35677-89-5 | |
| White | |
| MFCD00674555 | |
| N-Cbz-L-homoserine Lactone, (S)-alpha-(Carbobenzoxyamino)-gamma-butyrolactone, (S)-alpha-(Cbz-amino)-gamma-butyrolactone | |
| C1COC(=O)C1NC(=O)OCC2=CC=CC=C2 | |
| 235.239 | |
| 235.24 | |
| Crystalline Powder |
| 129°C | |
| C12H13NO4 | |
| 1 g | |
| FKWDZIFOVOUDAG-JTQLQIEISA-N | |
| benzyl N-[(3S)-2-oxooxolan-3-yl]carbamate | |
| 489185 | |
| ≥98.0% (HPLC,N) | |
| N-Carbobenzoxy-L-homoserine Lactone |
Safety and Handling
TSCA : No