missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-L-alaninol 98.0+%, TCI America™

Catalog No. C26295G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26295G 5 g
C26291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26295G Supplier TCI America Supplier No. C26295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66674-16-6
Molecular Formula C11H15NO3
Molecular Weight (g/mol) 209.25
MDL Number MFCD00672530
InChI Key AFPHMHSLDRPUSM-VIFPVBQESA-N
Synonym cbz-l-alaninol, s-benzyl 1-hydroxypropan-2-yl carbamate, n-z-l-alaninol, s-benzyl 1-hydroxypropan-2-ylcarbamate, benzyl n-2s-1-hydroxypropan-2-yl carbamate, carbamic acid, 1s-2-hydroxy-1-methylethyl-, phenylmethyl ester, phenylmethyl n-2s-1-oxidanylpropan-2-yl carbamate, n-2s-1-hydroxypropan-2-yl carbamic acid phenylmethyl ester, n-cbz-l-alaninol, cbz-l-ala-ol
PubChem CID 10262499
IUPAC Name benzyl N-[(2S)-1-hydroxypropan-2-yl]carbamate
SMILES C[C@@H](CO)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 83°C
Color White
Quantity 5 g
Formula Weight 209.25
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-alaninol