missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-L-alanine 99.0+%, TCI America™

Catalog No. C063325G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C063325G 25 g
C063310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C063325G Supplier TCI America Supplier No. C063325G

Chemical Identifiers

CAS 1142-20-7
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
MDL Number MFCD00002640
InChI Key TYRGLVWXHJRKMT-QMMMGPOBSA-N
Synonym n-carbobenzyloxy-l-alanine, z-ala-oh, n-cbz-l-alanine, z-l-alanine, n-benzyloxycarbonyl-l-alanine, cbz-l-alanine, cbz-ala-oh, carbobenzyloxy-l-alanine, benzyloxycarbonyl-l-alanine, carbobenzoxy-l-alanine
PubChem CID 736104
IUPAC Name (2S)-2-{[(benzyloxy)carbonyl]amino}propanoic acid
SMILES C[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 86°C
Color White
Quantity 25 g
Formula Weight 223.23
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-alanine