missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-DL-methionine 98.0+%, TCI America™
$29.83 - $136.08
Chemical Identifiers
| CAS | 4434-61-1 |
|---|---|
| Molecular Formula | C13H17NO4S |
| Molecular Weight (g/mol) | 283.342 |
| MDL Number | MFCD00065123 |
| InChI Key | FPKHNNQXKZMOJJ-UHFFFAOYSA-N |
| Synonym | N-Cbz-DL-methionine, Z-DL-Met-OH |
| PubChem CID | 97806 |
| IUPAC Name | 4-methylsulfanyl-2-(phenylmethoxycarbonylamino)butanoic acid |
| SMILES | CSCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1 |
Chemical Identifiers
| 4434-61-1 | |
| 283.342 | |
| FPKHNNQXKZMOJJ-UHFFFAOYSA-N | |
| 97806 | |
| CSCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1 |
| C13H17NO4S | |
| MFCD00065123 | |
| N-Cbz-DL-methionine, Z-DL-Met-OH | |
| 4-methylsulfanyl-2-(phenylmethoxycarbonylamino)butanoic acid |
Specifications
| 4434-61-1 | |
| White | |
| MFCD00065123 | |
| N-Cbz-DL-methionine, Z-DL-Met-OH | |
| CSCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1 | |
| 283.342 | |
| 283.34 | |
| Crystalline Powder |
| 113°C | |
| C13H17NO4S | |
| 1 g | |
| FPKHNNQXKZMOJJ-UHFFFAOYSA-N | |
| 4-methylsulfanyl-2-(phenylmethoxycarbonylamino)butanoic acid | |
| 97806 | |
| ≥98.0% (HPLC,T) | |
| N-Carbobenzoxy-DL-methionine |
Safety and Handling
TSCA : No