missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-D-phenylalanine 98.0+%, TCI America™

Catalog No. p-7124553
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C06621G 1 g
C06625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C06621G Supplier TCI America Supplier No. C06621G
Only null left

Chemical Identifiers

CAS 2448-45-5
Molecular Formula C17H17NO4
Molecular Weight (g/mol) 299.326
MDL Number MFCD00063151
InChI Key RRONHWAVOYADJL-OAHLLOKOSA-N
Synonym z-d-phe-oh, n-cbz-d-phenylalanine, z-d-phenylalanine, cbz-d-phe-oh, n-carbobenzyloxy-d-phenylalanine, chembl63865, benzyloxycarbonyl-d-phenylalanine, n-benzyloxycarbonyl-d-phenylalanine, 2r-3-phenyl-2-phenylmethoxycarbonylamino propanoic acid, d-phenylalanine, n-phenylmethoxy carbonyl
PubChem CID 675678
IUPAC Name (2R)-3-phenyl-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2=CC=CC=C2

Specifications

Melting Point 88°C
Color White
Quantity 1 g
Formula Weight 299.33
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-D-phenylalanine