missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-D-leucine Dicyclohexylammonium Salt 98.0+%, TCI America™
$54.06 - $162.17
Chemical Identifiers
| CAS | 7662-58-0 |
|---|---|
| Molecular Formula | C26H42N2O4 |
| Molecular Weight (g/mol) | 446.63 |
| MDL Number | MFCD10566885 |
| InChI Key | FOULZFSGIVQTHX-HVPILOLGNA-N |
| Synonym | z-d-leu-oh.dcha, n-carbobenzoxy-d-leucine dicyclohexylammonium salt, dicyclohexylamine r-2-benzyloxy carbonyl amino-4-methylpentanoate, 2r-2-benzyloxy carbonyl amino-4-methylpentanoic acid; dicha, z-d-leu-ohcha, n-z-leu-oh.dcha, n-cbz-d-leucine dicyclohexylammonium salt, n-benzyloxycarbonyl-d-leucine.dicyclohexylamine, n-carbobenzoxy-d-leucinedicyclohexylammoniumsalt, n-benzyloxycarbonyl-d-leucine dicyclohexylammonium salt |
| PubChem CID | 44629899 |
| IUPAC Name | (2R)-2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoic acid; N-cyclohexylcyclohexanamine |
| SMILES | C1CCC(CC1)NC1CCCCC1.CC(C)C[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O |
Chemical Identifiers
| 7662-58-0 | |
| 446.63 | |
| FOULZFSGIVQTHX-HVPILOLGNA-N | |
| 44629899 | |
| C1CCC(CC1)NC1CCCCC1.CC(C)C[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O |
| C26H42N2O4 | |
| MFCD10566885 | |
| z-d-leu-oh.dcha, n-carbobenzoxy-d-leucine dicyclohexylammonium salt, dicyclohexylamine r-2-benzyloxy carbonyl amino-4-methylpentanoate, 2r-2-benzyloxy carbonyl amino-4-methylpentanoic acid; dicha, z-d-leu-ohcha, n-z-leu-oh.dcha, n-cbz-d-leucine dicyclohexylammonium salt, n-benzyloxycarbonyl-d-leucine.dicyclohexylamine, n-carbobenzoxy-d-leucinedicyclohexylammoniumsalt, n-benzyloxycarbonyl-d-leucine dicyclohexylammonium salt | |
| (2R)-2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoic acid; N-cyclohexylcyclohexanamine |
Specifications
| 7662-58-0 | |
| White | |
| MFCD10566885 | |
| z-d-leu-oh.dcha, n-carbobenzoxy-d-leucine dicyclohexylammonium salt, dicyclohexylamine r-2-benzyloxy carbonyl amino-4-methylpentanoate, 2r-2-benzyloxy carbonyl amino-4-methylpentanoic acid; dicha, z-d-leu-ohcha, n-z-leu-oh.dcha, n-cbz-d-leucine dicyclohexylammonium salt, n-benzyloxycarbonyl-d-leucine.dicyclohexylamine, n-carbobenzoxy-d-leucinedicyclohexylammoniumsalt, n-benzyloxycarbonyl-d-leucine dicyclohexylammonium salt | |
| C1CCC(CC1)NC1CCCCC1.CC(C)C[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O | |
| 446.63 | |
| 446.63 | |
| Crystalline Powder |
| 154°C | |
| C26H42N2O4 | |
| 1 g | |
| FOULZFSGIVQTHX-HVPILOLGNA-N | |
| (2R)-2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoic acid; N-cyclohexylcyclohexanamine | |
| 44629899 | |
| ≥98.0% (T) | |
| N-Carbobenzoxy-D-leucine Dicyclohexylammonium Salt |
Safety and Handling
TSCA : No