missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-D-glutamic Acid 98.0+%, TCI America™
$61.51 - $182.68
Chemical Identifiers
| CAS | 63648-73-7 |
|---|---|
| Molecular Formula | C13H15NO6 |
| Molecular Weight (g/mol) | 281.264 |
| MDL Number | MFCD00063193 |
| InChI Key | PVFCXMDXBIEMQG-SNVBAGLBSA-N |
| Synonym | z-d-glu-oh, z-d-glutamic acid, n-carbobenzoxy-d-glutamic acid, z-d-glu, n-cbz-d-glutamic acid, n-benzyloxycarbonyl-d-glutamic acid, r-2-benzyloxy carbonyl amino pentanedioic acid, cbz-glu-oh, 2r-2-benzyloxy carbonyl amino pentanedioic acid, cbz-d-glu |
| PubChem CID | 1607568 |
| IUPAC Name | (2R)-2-(phenylmethoxycarbonylamino)pentanedioic acid |
| SMILES | C1=CC=C(C=C1)COC(=O)NC(CCC(=O)O)C(=O)O |
Chemical Identifiers
| 63648-73-7 | |
| 281.264 | |
| PVFCXMDXBIEMQG-SNVBAGLBSA-N | |
| 1607568 | |
| C1=CC=C(C=C1)COC(=O)NC(CCC(=O)O)C(=O)O |
| C13H15NO6 | |
| MFCD00063193 | |
| z-d-glu-oh, z-d-glutamic acid, n-carbobenzoxy-d-glutamic acid, z-d-glu, n-cbz-d-glutamic acid, n-benzyloxycarbonyl-d-glutamic acid, r-2-benzyloxy carbonyl amino pentanedioic acid, cbz-glu-oh, 2r-2-benzyloxy carbonyl amino pentanedioic acid, cbz-d-glu | |
| (2R)-2-(phenylmethoxycarbonylamino)pentanedioic acid |
Specifications
| 63648-73-7 | |
| White | |
| MFCD00063193 | |
| z-d-glu-oh, z-d-glutamic acid, n-carbobenzoxy-d-glutamic acid, z-d-glu, n-cbz-d-glutamic acid, n-benzyloxycarbonyl-d-glutamic acid, r-2-benzyloxy carbonyl amino pentanedioic acid, cbz-glu-oh, 2r-2-benzyloxy carbonyl amino pentanedioic acid, cbz-d-glu | |
| C1=CC=C(C=C1)COC(=O)NC(CCC(=O)O)C(=O)O | |
| 281.264 | |
| 281.26 | |
| Crystalline Powder |
| 120°C | |
| C13H15NO6 | |
| 5 g | |
| PVFCXMDXBIEMQG-SNVBAGLBSA-N | |
| (2R)-2-(phenylmethoxycarbonylamino)pentanedioic acid | |
| 1607568 | |
| ≥98.0% (T) | |
| N-Carbobenzoxy-D-glutamic Acid |
Safety and Handling
TSCA : No