missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-D-aspartic Acid 98.0+%, TCI America™

Catalog No. C06891G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C06891G 1 g
C06895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C06891G Supplier TCI America Supplier No. C06891G
Only null left

Chemical Identifiers

CAS 78663-07-7
Molecular Formula C12H13NO6
Molecular Weight (g/mol) 267.237
MDL Number MFCD00063182
InChI Key XYXYXSKSTZAEJW-SECBINFHSA-N
Synonym z-d-asp-oh, n-benzyloxycarbonyl-d-aspartic acid, z-d-aspartic acid, cbz-d-aspartic acid, n-cbz-d-aspartic acid, r-2-benzyloxy carbonyl amino succinic acid, cbz-d-asp-oh, n-carbobenzoxy-d-aspartic acid, r-2-benzyloxycarbonylamino succinic acid, 2r-2-benzyloxy carbonyl amino butanedioic acid
PubChem CID 5289152
IUPAC Name (2R)-2-(phenylmethoxycarbonylamino)butanedioic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CC(=O)O)C(=O)O

Specifications

Melting Point 117°C
Color White
Quantity 1 g
Formula Weight 267.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-D-aspartic Acid
EINECSNumber (9)-1549
TSCA No