missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-D-2-phenylglycine 98.0+%, TCI America™

Catalog No. p-7125531
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25661G 1 g
C25665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25661G Supplier TCI America Supplier No. C25661G
Only null left

Chemical Identifiers

CAS 17609-52-8
Molecular Formula C16H15NO4
Molecular Weight (g/mol) 285.299
MDL Number MFCD00021703
InChI Key RLDJWBVOZVJJOS-CQSZACIVSA-N
Synonym z-d-phg-oh, cbz-d---phenylglycine, r-2-benzyloxy carbonyl amino-2-phenylacetic acid, z-d-phenylglycine, z-d-2-phenylglycine, n-carbobenzoxy-d-2-phenylglycine, 2r-benzyloxy carbonyl amino phenyl acetic acid, cbz-d-phenylglycine, r-n-benzyloxycarbonyl-alpha-carboxybenzylamine, n-alpha-benzyloxycarbonyl-d-phenylglycine
PubChem CID 87183
IUPAC Name (2R)-2-phenyl-2-(phenylmethoxycarbonylamino)acetic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 131°C
Color White
Quantity 1 g
Formula Weight 285.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-D-2-phenylglycine