missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-4-hydroxy-D-2-phenylglycine 98.0+%, TCI America™

Catalog No. C27735G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27735G 5 g
C277325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27735G Supplier TCI America Supplier No. C27735G
Only null left

Chemical Identifiers

CAS 26787-75-7
Molecular Formula C16H15NO5
Molecular Weight (g/mol) 301.30
MDL Number MFCD09756650
InChI Key CGDQNVNWDDEJPY-UHFFFAOYNA-N
Synonym (R)-2-(Carbobenzoxy)amino-2-(4-hydroxyphenyl)acetic Acid, Z-D-Phg(4-OH)-OH
PubChem CID 1514128
IUPAC Name 2-{[(benzyloxy)carbonyl]amino}-2-(4-hydroxyphenyl)acetic acid
SMILES OC(=O)C(NC(=O)OCC1=CC=CC=C1)C1=CC=C(O)C=C1

Specifications

Color White
Quantity 5 g
Formula Weight 301.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-4-hydroxy-D-2-phenylglycine