missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-4-aminobutyric Acid 98.0+%, TCI America™

Catalog No. p-7124590
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07531G 1 g
C075325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07531G Supplier TCI America Supplier No. C07531G
Only null left

Chemical Identifiers

CAS 5105-78-2
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.26
MDL Number MFCD00055835
InChI Key STQMDRQJSNKUAW-UHFFFAOYSA-N
Synonym z-gamma-abu-oh, 4-benzyloxy carbonyl amino butanoic acid, n-cbz-gaba, carbobenzoxy-4-aminobutyric acid, n-cbz-4-aminobutanoic acid, z-gaba-oh, n-cbz-4-aminobutyric acid, 4-carboxyamino butyric acid n-benzyl ester, z-l-gamma-aminobutyric acid
PubChem CID 21184
IUPAC Name 4-{[(benzyloxy)carbonyl]amino}butanoic acid
SMILES OC(=O)CCCNC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 64°C
Color White
Quantity 1 g
Formula Weight 237.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-4-aminobutyric Acid
RTECSNumber ES8137200
TSCA No