missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-4-aminobutyric Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 64°C |
| Color | White |
| Quantity | 25 g |
| Formula Weight | 237.26 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-Carbobenzoxy-4-aminobutyric Acid |
Safety and Handling
| RTECSNumber | ES8137200 |
| TSCA | No |