missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-2-phosphonoglycine Trimethyl Ester 98.0+%, TCI America™
$96.94 - $201.32
Chemical Identifiers
| CAS | 88568-95-0 |
|---|---|
| Molecular Formula | C13H18NO7P |
| Molecular Weight (g/mol) | 331.261 |
| MDL Number | MFCD00043304 |
| InChI Key | GSYSFVSGPABNNL-UHFFFAOYSA-N |
| Synonym | methyl 2-benzyloxy carbonyl amino-2-dimethoxyphosphoryl acetate, +/--benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester, n-cbz-2-phosphonoglycine trimethyl ester, n-benzyloxy carbonyl, +/--cbz-alpha-phosphonoglycine trimethyl ester, methyl 2-benzyloxycarbonylamino-2-dimethoxyphosphoryl-acetate, +/--n-cbz-alpha-phosphonoglycine trimethyl ester, z-alpha-phosphonoglycine trimethyl ester, cbz-alpha-phosphono-dl-glycine trimethyl ester |
| PubChem CID | 2734718 |
| IUPAC Name | methyl 2-dimethoxyphosphoryl-2-(phenylmethoxycarbonylamino)acetate |
| SMILES | COC(=O)C(NC(=O)OCC1=CC=CC=C1)P(=O)(OC)OC |
Chemical Identifiers
| 88568-95-0 | |
| 331.261 | |
| GSYSFVSGPABNNL-UHFFFAOYSA-N | |
| 2734718 | |
| COC(=O)C(NC(=O)OCC1=CC=CC=C1)P(=O)(OC)OC |
| C13H18NO7P | |
| MFCD00043304 | |
| methyl 2-benzyloxy carbonyl amino-2-dimethoxyphosphoryl acetate, +/--benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester, n-cbz-2-phosphonoglycine trimethyl ester, n-benzyloxy carbonyl, +/--cbz-alpha-phosphonoglycine trimethyl ester, methyl 2-benzyloxycarbonylamino-2-dimethoxyphosphoryl-acetate, +/--n-cbz-alpha-phosphonoglycine trimethyl ester, z-alpha-phosphonoglycine trimethyl ester, cbz-alpha-phosphono-dl-glycine trimethyl ester | |
| methyl 2-dimethoxyphosphoryl-2-(phenylmethoxycarbonylamino)acetate |
Specifications
| 88568-95-0 | |
| White-Yellow | |
| MFCD00043304 | |
| methyl 2-benzyloxy carbonyl amino-2-dimethoxyphosphoryl acetate, +/--benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester, n-cbz-2-phosphonoglycine trimethyl ester, n-benzyloxy carbonyl, +/--cbz-alpha-phosphonoglycine trimethyl ester, methyl 2-benzyloxycarbonylamino-2-dimethoxyphosphoryl-acetate, +/--n-cbz-alpha-phosphonoglycine trimethyl ester, z-alpha-phosphonoglycine trimethyl ester, cbz-alpha-phosphono-dl-glycine trimethyl ester | |
| COC(=O)C(NC(=O)OCC1=CC=CC=C1)P(=O)(OC)OC | |
| 331.261 | |
| 331.26 | |
| Crystalline Powder |
| 80°C | |
| C13H18NO7P | |
| 1 g | |
| GSYSFVSGPABNNL-UHFFFAOYSA-N | |
| methyl 2-dimethoxyphosphoryl-2-(phenylmethoxycarbonylamino)acetate | |
| 2734718 | |
| ≥98.0% (HPLC,N) | |
| N-Carbobenzoxy-2-phosphonoglycine Trimethyl Ester |
Safety and Handling
TSCA : No