missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-2-phosphonoglycine Trimethyl Ester 98.0+%, TCI America™

Catalog No. C24401G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24401G 1 g
C24405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24401G Supplier TCI America Supplier No. C24401G
Only null left

Chemical Identifiers

CAS 88568-95-0
Molecular Formula C13H18NO7P
Molecular Weight (g/mol) 331.261
MDL Number MFCD00043304
InChI Key GSYSFVSGPABNNL-UHFFFAOYSA-N
Synonym methyl 2-benzyloxy carbonyl amino-2-dimethoxyphosphoryl acetate, +/--benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester, n-cbz-2-phosphonoglycine trimethyl ester, n-benzyloxy carbonyl, +/--cbz-alpha-phosphonoglycine trimethyl ester, methyl 2-benzyloxycarbonylamino-2-dimethoxyphosphoryl-acetate, +/--n-cbz-alpha-phosphonoglycine trimethyl ester, z-alpha-phosphonoglycine trimethyl ester, cbz-alpha-phosphono-dl-glycine trimethyl ester
PubChem CID 2734718
IUPAC Name methyl 2-dimethoxyphosphoryl-2-(phenylmethoxycarbonylamino)acetate
SMILES COC(=O)C(NC(=O)OCC1=CC=CC=C1)P(=O)(OC)OC

Specifications

Melting Point 80°C
Color White-Yellow
Quantity 1 g
Formula Weight 331.26
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-2-phosphonoglycine Trimethyl Ester