missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-1,3-diaminopropane Hydrochloride 98.0+%, TCI America™
Supplier: TCI America C15125G
Specifications
| N-Carbobenzoxy-1,3-diaminopropane Hydrochloride | |
| 186°C | |
| C11H17ClN2O2 | |
| 5 g | |
| XKMBTMXQMDLSRB-UHFFFAOYSA-N | |
| hydrogen benzyl N-(3-aminopropyl)carbamate chloride | |
| 13196227 | |
| ≥98.0% (T) |
| 17400-34-9 | |
| White-Yellow | |
| MFCD00270153 | |
| n-cbz-1,3-diaminopropane hydrochloride, benzyl 3-aminopropyl carbamate hydrochloride, benzyl n-3-aminopropyl carbamate hydrochloride, n-cbz-1,3-diaminopropane-hcl, n-z-1,3-propanediamine hydrochloride, n-carbobenzoxy-1,3-diaminopropane hydrochloride, n-carbobenzyloxy-1,3-diaminopropane hydrochloride, n-3-aminopropyl carbamic acid benzyl ester hydrochloride, n-1-z-1,3-diaminopropane hcl, n-z-1,3-diaminopropane hydrochloride | |
| [H+].[Cl-].NCCCNC(=O)OCC1=CC=CC=C1 | |
| 244.72 | |
| 244.72 | |
| Crystalline Powder |
Chemical Identifiers
| 17400-34-9 | |
| 244.72 | |
| XKMBTMXQMDLSRB-UHFFFAOYSA-N | |
| 13196227 | |
| [H+].[Cl-].NCCCNC(=O)OCC1=CC=CC=C1 |
| C11H17ClN2O2 | |
| MFCD00270153 | |
| n-cbz-1,3-diaminopropane hydrochloride, benzyl 3-aminopropyl carbamate hydrochloride, benzyl n-3-aminopropyl carbamate hydrochloride, n-cbz-1,3-diaminopropane-hcl, n-z-1,3-propanediamine hydrochloride, n-carbobenzoxy-1,3-diaminopropane hydrochloride, n-carbobenzyloxy-1,3-diaminopropane hydrochloride, n-3-aminopropyl carbamic acid benzyl ester hydrochloride, n-1-z-1,3-diaminopropane hcl, n-z-1,3-diaminopropane hydrochloride | |
| hydrogen benzyl N-(3-aminopropyl)carbamate chloride |
Safety and Handling
TSCA : No