missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-1,3-diaminopropane Hydrochloride 98.0+%, TCI America™

Catalog No. C15121G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15121G 1 g
C15125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15121G Supplier TCI America Supplier No. C15121G

Chemical Identifiers

CAS 17400-34-9
Molecular Formula C11H17ClN2O2
Molecular Weight (g/mol) 244.72
MDL Number MFCD00270153
InChI Key XKMBTMXQMDLSRB-UHFFFAOYSA-N
Synonym n-cbz-1,3-diaminopropane hydrochloride, benzyl 3-aminopropyl carbamate hydrochloride, benzyl n-3-aminopropyl carbamate hydrochloride, n-cbz-1,3-diaminopropane-hcl, n-z-1,3-propanediamine hydrochloride, n-carbobenzoxy-1,3-diaminopropane hydrochloride, n-carbobenzyloxy-1,3-diaminopropane hydrochloride, n-3-aminopropyl carbamic acid benzyl ester hydrochloride, n-1-z-1,3-diaminopropane hcl, n-z-1,3-diaminopropane hydrochloride
PubChem CID 13196227
IUPAC Name hydrogen benzyl N-(3-aminopropyl)carbamate chloride
SMILES [H+].[Cl-].NCCCNC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 186°C
Color White-Yellow
Quantity 1 g
Formula Weight 244.72
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-1,3-diaminopropane Hydrochloride