missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-1,2-diaminoethane Hydrochloride 98.0+%, TCI America™

Catalog No. C15111G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15111G 1 g
C15115G 5 g
C151125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. C15111G missing translation for 'mfr' TCI America missing translation for 'supplierNo' C15111G

Chemical Identifiers

CAS 18807-71-1
Molecular Formula C10H15ClN2O2
Molecular Weight (g/mol) 230.69
MDL Number MFCD00270150
InChI Key QMLKQXIAPAAIEJ-UHFFFAOYSA-N
Synonym benzyl n-2-aminoethyl carbamate hydrochloride, n-cbz-ethylenediamine hydrochloride, benzyl 2-aminoethyl carbamate hydrochloride, n-carbobenzoxy-1,2-diaminoethane hydrochloride, n-z-ethylenediamine hydrochloride, n-benzyloxycarbonyl-1,2-diaminoethane hydrochloride, n-z-1,2-diaminoethane hydrochloride, n-cbz-1,2-diaminoethane hydrochloride, benzyl 2-aminoethyl carbamate hcl, n-benzyloxycarbonylethylenediamine hydrochloride
PubChem CID 12715871
IUPAC Name benzyl N-(2-aminoethyl)carbamate hydrochloride
SMILES Cl.NCCNC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 172°C
Quantity 1 g
Formula Weight 230.69
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-1,2-diaminoethane Hydrochloride