missing translation for 'onlineSavingsMsg'
Learn More

N-Carbethoxyphthalimide, 99+%

Catalog No. AC108310500
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108310500 50 g Glass bottle
AC108312500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC108310500 Supplier Thermo Scientific Chemicals Supplier No. 108310500

CAS: 22509-74-6 | C11H9NO4 | 219.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22509-74-6
Molecular Formula C11H9NO4
Molecular Weight (g/mol) 219.2
MDL Number MFCD00005893
InChI Key VRHAQNTWKSVEEC-UHFFFAOYSA-N
Synonym n-carbethoxyphthalimide, n-ethoxycarbonylphthalimide, n-carboethoxyphthalimide, phthalimide-n-carbethoxy, ethyl n-phthaloylcarbamate, n-ethoxycarbonyl phthalimide, 2h-isoindole-2-carboxylic acid, 1,3-dihydro-1,3-dioxo-, ethyl ester, ethyl 1,3-dioxoisoindoline-2-carboxylate, n-karbetoksi-ftalimid, 2-isoindolinecarboxylic acid, 1,3-dioxo-, ethyl ester
PubChem CID 31187
IUPAC Name ethyl 1,3-dioxoisoindole-2-carboxylate
SMILES CCOC(=O)N1C(=O)C2=CC=CC=C2C1=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 82.0°C to 84.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 50 g
Beilstein 21, II, 357
Fieser 01,111; 05,91; 09,212
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform
Formula Weight 219.2
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material N-Carbethoxyphthalimide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 245-048-5

RUO – Research Use Only