missing translation for 'onlineSavingsMsg'
Learn More

N-Carbethoxy-4-piperidone, 98.5+%

CAS: 29976-53-2 | C8H13NO3 | 171.2 g/mol

$75.05 - $75.05

Chemical Identifiers

CAS 29976-53-2
Molecular Formula C8H13NO3
Molecular Weight (g/mol) 171.2
MDL Number MFCD00006188
InChI Key LUBGFMZTGFXIIN-UHFFFAOYSA-N
Synonym n-carbethoxy-4-piperidone, ethyl 4-oxo-1-piperidinecarboxylate, n-carboethoxy-4-piperidone, 1-ethoxycarbonyl-4-piperidone, 1-carbethoxy-4-piperidone, 4-oxo-1-piperidinecarboxylic acid ethyl ester, n-ethoxycarbonyl-4-piperidone, unii-7h5jx1s259, 1-piperidinecarboxylic acid, 4-oxo-, ethyl ester, ethyl 4-oxopiperidinecarboxylate
PubChem CID 34767
IUPAC Name ethyl 4-oxopiperidine-1-carboxylate
SMILES CCOC(=O)N1CCC(=O)CC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC167240100
SDS View Documents
Thermo Scientific Chemicals
167240100
10 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

29976-53-2
171.2
LUBGFMZTGFXIIN-UHFFFAOYSA-N
34767
CCOC(=O)N1CCC(=O)CC1
C8H13NO3
MFCD00006188
n-carbethoxy-4-piperidone, ethyl 4-oxo-1-piperidinecarboxylate, n-carboethoxy-4-piperidone, 1-ethoxycarbonyl-4-piperidone, 1-carbethoxy-4-piperidone, 4-oxo-1-piperidinecarboxylic acid ethyl ester, n-ethoxycarbonyl-4-piperidone, unii-7h5jx1s259, 1-piperidinecarboxylic acid, 4-oxo-, ethyl ester, ethyl 4-oxopiperidinecarboxylate
ethyl 4-oxopiperidine-1-carboxylate

Specifications

29976-53-2
100.0
1.1350g/mL
87°C
98.5% min. (GC)
C8H13NO3
MFCD00006188
1.135
Solubility in water: slightly soluble
CCOC(=O)N1CCC(=O)CC1
171.2
171.2
Liquid
98.5
Colorless to Yellow
95.0°C to 98.0°C (1.0 mmHg)
Authentic
Glass bottle
1.4740 to 1.4760
10 g
n-carbethoxy-4-piperidone, ethyl 4-oxo-1-piperidinecarboxylate, n-carboethoxy-4-piperidone, 1-ethoxycarbonyl-4-piperidone, 1-carbethoxy-4-piperidone, 4-oxo-1-piperidinecarboxylic acid ethyl ester, n-ethoxycarbonyl-4-piperidone, unii-7h5jx1s259, 1-piperidinecarboxylic acid, 4-oxo-, ethyl ester, ethyl 4-oxopiperidinecarboxylate
LUBGFMZTGFXIIN-UHFFFAOYSA-N
ethyl 4-oxopiperidine-1-carboxylate
34767
98.5+%
N-Carbethoxy-4-piperidone
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 249-984-5

Recommended Storage : Product may darken during storage

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only