missing translation for 'onlineSavingsMsg'
Learn More

n-Butyric acid, 99+%

CAS: 107-92-6 | C4H8O2 | 88.11 g/mol

$61.96 - $280.23

Chemical Identifiers

CAS 107-92-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00002814
InChI Key FERIUCNNQQJTOY-UHFFFAOYSA-N
Synonym butyric acid, n-butyric acid, n-butanoic acid, propylformic acid, ethylacetic acid, butanic acid, 1-propanecarboxylic acid, butyrate, 1-butyric acid, buttersaeure
PubChem CID 264
ChEBI CHEBI:30772
IUPAC Name butanoic acid
SMILES CCCC(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC108111000
SDS View Documents
Thermo Scientific Chemicals
108111000
100 mL Glass bottle
Each for $61.96
Only null left
 
AC108110010
SDS View Documents
Thermo Scientific Chemicals
108110010
1 L Glass bottle
Each for $124.19
Only null left
 
AC108110025
SDS View Documents
Thermo Scientific Chemicals
108110025
2.5 L Glass bottle
Each for $280.23
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

107-92-6
88.11
FERIUCNNQQJTOY-UHFFFAOYSA-N
264
butanoic acid
C4H8O2
MFCD00002814
butyric acid, n-butyric acid, n-butanoic acid, propylformic acid, ethylacetic acid, butanic acid, 1-propanecarboxylic acid, butyrate, 1-butyric acid, buttersaeure
CHEBI:30772
CCCC(O)=O

Specifications

-7.0°C to +5.0°C
162.0°C to 165.0°C
100.0
C4H8O2
MFCD00002814
butyric acid, n-butyric acid, n-butanoic acid, propylformic acid, ethylacetic acid, butanic acid, 1-propanecarboxylic acid, butyrate, 1-butyric acid, buttersaeure
FERIUCNNQQJTOY-UHFFFAOYSA-N
butanoic acid
264
88.11
0.9600g/mL
69°C
Glass bottle
1.3965 to 1.3995
n-Butyric acid
107-92-6
99.0
99+%
CH3CH2CH2CO2H
2820
Solubility in water: miscible. Other solubilities: miscible with almost all common organic solvents
CCCC(O)=O
88.11
CHEBI:30772
≥99%
100 mL
Authentic
Liquid
0.96
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
Documents

Documents

RUO – Research Use Only