missing translation for 'onlineSavingsMsg'
Learn More

n-Butyric Acid, 98%, Spectrum™ Chemical

Catalog No. 18611913
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Poly-Coated Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611913 100 mL Amber Glass Bottle
18611912 1 L Amber Glass Bottle
18611914 2.5 L Poly-Coated Glass Bottle
18611915 20 L Poly Drum
18611916 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18611913 Supplier Spectrum Chemical Mfg Cor Supplier No. B1195100ML
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 107-92-6

Specifications

CAS Max % 1
Percent Purity 98%
Grade Reagent
Quantity 100 mL
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.