missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Butyric Acid, 98%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Percent Purity | 98% |
| Grade | Reagent |
| Quantity | 1 L |
| Packaging | Amber Glass Bottle |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.